missing translation for 'onlineSavingsMsg'
Learn More

Carnauba Wax, Yellow No. 1, Flakes, Spectrum™ Chemical

Catalog No. 18609587
Change view
Click to view available options
Quantity:
500 g
2.5 kg
25 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18609587 500 g Poly Bottle
18609588 2.5 kg Poly Pail
18609589 25 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18609587 Supplier Spectrum Chemical Mfg Cor Supplier No. CA196500GM

Chemical Identifiers

CAS 8015-86-9

Specifications

CAS Max % 1
Melting Range 82.0° to 85.5°C
Packaging Poly Bottle
Quantity 500 g
Grade Reagent

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.