missing translation for 'onlineSavingsMsg'
Learn More

Carnauba wax, refined, No. 1, yellow

CAS: 8015-86-9

Supplier:  Thermo Scientific Chemicals 222380010

 View more versions of this product

Catalog No. AC222380010


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Carnauba wax
8015-86-9
Yellow
0.25% max.
MFCD00130724
78 to 95mg KOH/g
Brazil wax
Powder or Flakes
2 to 7 (mg KOH/g)
81.0°C to 86.0°C
282°C
Plastic bottle
1 kg
15,1846
Solubility in water: insoluble. Other solubilities: soluble in boiling alcohol,diethyl ether,benzene,,toluene,chloroform,ethyl acetate,turpentine,,petroleum ether,carbon disulfide;,soluble in alkalies

Chemical Identifiers

8015-86-9
MFCD00130724
Brazil wax
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 232-399-4

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only