missing translation for 'onlineSavingsMsg'
Learn More

Carmine Reagent, for Boron determination, Ricca Chemical

Catalog No. 185016
Change view
Click to view available options
Package: Safety-Coated Glass; Size: 500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
185016 Package: Safety-Coated Glass; Size: 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 185016 Supplier Ricca Chemical Company Supplier No. RI185016

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For Boron determination

Specifications

Chemical Name or Material Carmine Reagent
Name Note Carmine Reagent
CAS 1390-65-4, 7664-93-9
CAS Min % 98.0, 0.05
CAS Max % 100.0, 0.05
Packaging Safety-coated Glass Bottle
Quantity 500 mL