missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Carmine, pure, high purity biol.stain, alum lake of carminic acid

Catalog No. AC190200050
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC190200050 5 g Glass bottle
AC190200250 25 g Glass bottle
AC190201000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC190200050 Supplier Thermo Scientific Chemicals Supplier No. 190200050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1390-65-4 | C22H20O13 | 492.39 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1390-65-4
Molecular Formula C22H20O13
Molecular Weight (g/mol) 492.39
MDL Number MFCD00167028
InChI Key DGQLVPJVXFOQEV-UHFFFAOYNA-N
Synonym C.I. 75470
PubChem CID 25113282
IUPAC Name 3,5,6,8-tetrahydroxy-1-methyl-9,10-dioxo-7-[(2S,3R,4S,5R,6S)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]anthracene-2-carboxylic acid
SMILES CC1=C(C(O)=O)C(O)=CC2=C1C(=O)C1=C(O)C(C3OC(CO)C(O)C(O)C3O)=C(O)C(O)=C1C2=O

Specifications

Infrared Spectrum Authentic
Loss on Drying 20% max.
Packaging Glass bottle
UV Spectrometry Complies
Merck Index 15, 2442
Color Purple/Red to Red
Solubility Information Solubility in water: soluble in hot water. Other solubilities: soluble in alkali, insoluble in diluted acids
Quantity 5 g
Physical Form Powder
Percent Purity ≥42.0% (carminic acid)
Grade Pure
Chemical Name or Material Carmine
Show More Show Less

RUO – Research Use Only