missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Carisoprodol

Supplier:  Thermo Scientific™ 466240010

Quantity 1 g

Catalog No. AC466240010



Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

78-44-4
Carisoprodol

Specifications

Carisoprodol
1 g
90.0°C to 94.0°C
78-44-4
Solubility aq. soln.: .. Other solubilities: soluble in dmso, chloroform, methanol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement Harmful if swallowed.

GHS P Statement IF SWALLOWED: rinse mouth.

Do NOT induce vomiting.

Call a POISON CENTER or doctor/physician if you feel unwell.

Wash face, hands and any exposed skin thoroughly after handling.

GHS Signal Word: Warning

EINECSNumber : 201-118-7

SDS
Documents

Documents