missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cargille Laboratories Refractive Index Liquid Reference Standards
Description
Includes
- Data sheet
- Each single set contains the number of liquids specified in the chart
- Temperature-coefficient and dispersion-value data printed on label of each vial
- Each liquid is in a 7.4cc glass vial with plastic screw cap and glass applicator rod; packed in partitioned box
Order Info
Test materials may be ordered separately. Technical Bulletins are available on request.
†Certified Set.
‡Specially selected for required high-dispersion characteristics needed in dispersion staining, focal masking and double-variation refractometry.
Specifications
Specifications
| Type | Refractive Index Standards Set AA-1 |
| Refractive Index | 1.4 to 1.45 |
| For Use With (Equipment) | Refractometer |
| Accuracy | ±0.0002 |
| Increments | 0.002 |
| Includes | Data sheet, 30 standards |