missing translation for 'onlineSavingsMsg'
Learn More

Cargille Laboratories Refractive Index Liquid Reference Standards

Catalog No. 1394614C
Change view
Click to view available options
Accuracy:
±0.0002
±0.0005
Increments:
0.002
0.01
Refractive Index:
1.4 to 1.45
1.71 to 1.8
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Accuracy Increments Refractive Index
1394614C ±0.0005 0.01 1.71 to 1.8
1394611A ±0.0002 0.002 1.4 to 1.45
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 1394614C Supplier Cargille Supplier No. 18152C

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

  • Data sheet

Choose from seven single sets of Refractive Index liquids

  • Each single set contains the number of liquids specified in the chart
  • Temperature-coefficient and dispersion-value data printed on label of each vial
  • Each liquid is in a 7.4cc glass vial with plastic screw cap and glass applicator rod; packed in partitioned box

Order Info

Test materials may be ordered separately. Technical Bulletins are available on request.

†Certified Set.

‡Specially selected for required high-dispersion characteristics needed in dispersion staining, focal masking and double-variation refractometry.

Specifications

Type Refractive Index Standards Set M-1/2
Refractive Index 1.71 to 1.8
For Use With (Equipment) Refractometer
Accuracy ±0.0005
Increments 0.01
Includes Data sheet, 10 standards