Learn More
Foxx Life Sciences EZLabpure&; 20L Round Polypropylene Carboy, 83B (83mm) VersaCap Closure with Spigot
GREENER_CHOICE
Catalog No. NC1818184
Encompass
Foxx Life Sciences offers a premium line of high quality, Round Polypropylene (PP) laboratory Carboys comes with patented VersaCap technology. VersaCap offers large handles with finger grips that allow for easy tightening and opening. Our versatile leak proof carboys with strong easy grip handles, aim to take full advantage of storage efficiency at minimal effort. Overall functionality is available for many applications, in 10 Liter or 20 Liter sizes, with or without spigots each with a VersaCap 83B Polypropylene (PP) Cap. 20 Liter Carboy comes with Graduation marks.Foxx laboratory Polypropylene (PP) carboys are made from a USP Class VI, FDA Grade, autoclavable and Heavy Metal Free material. The recommended autoclaving cycle is: 15 PSI for 20 minutes at 121C (250F).
Catalog No. NC1818184 Supplier Foxx Life Sciences Supplier No. 155C817FLS
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Foxx Life Sciences offers a premium line of high quality, Round Polypropylene (PP) laboratory Carboys comes with patented VersaCap technology. VersaCap offers large handles with finger grips that allow for easy tightening and opening. Our versatile leak proof carboys with strong easy grip handles, aim to take full advantage of storage efficiency at minimal effort. Overall functionality is available for many applications, in 10 Liter or 20 Liter sizes, with or without spigots each with a VersaCap 83B Polypropylene (PP) Cap. 20 Liter Carboy comes with Graduation marks.Foxx laboratory Polypropylene (PP) carboys are made from a USP Class VI, FDA Grade, autoclavable and Heavy Metal Free material. The recommended autoclaving cycle is: 15 PSI for 20 minutes at 121C (250F).