Learn More
Foxx Life Sciences Wide Mouth 10L Round Polypropylene (PP) Carboy with White Cap
Catalog No. NC1818185
Encompass
Foxx Life Sciences offers a premium line of high quality, Round Wide Mouth Polypropylene (PP) Laboratory Carboys. Our versatile carboys with strong easy grip handles aim to take full advantage of storage. The Wide Mouth carboys are leak proof and allows for easy cleaning efficiency. Overall functionality is available in 10 Liter or 20 Liter sizes, without spigots each with a White HDPE material Cap. The Wide Mouth Carboy are Autoclavable. Foxx laboratory Polypropylene (PP) carboys are made from a USP Class VI, FDA Grade, autoclavable and Heavy Metal Free material. The recommended autoclaving cycle is: 15 PSI for 20 minutes at 121C (250F).Cap should be removed before Autoclaving this carboy.
Catalog No. NC1818185 Supplier Foxx Life Sciences Supplier No. 155B187FLS
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Foxx Life Sciences offers a premium line of high quality, Round Wide Mouth Polypropylene (PP) Laboratory Carboys. Our versatile carboys with strong easy grip handles aim to take full advantage of storage. The Wide Mouth carboys are leak proof and allows for easy cleaning efficiency. Overall functionality is available in 10 Liter or 20 Liter sizes, without spigots each with a White HDPE material Cap. The Wide Mouth Carboy are Autoclavable. Foxx laboratory Polypropylene (PP) carboys are made from a USP Class VI, FDA Grade, autoclavable and Heavy Metal Free material. The recommended autoclaving cycle is: 15 PSI for 20 minutes at 121C (250F).Cap should be removed before Autoclaving this carboy.