Learn More
Foxx Life Sciences EZLabpure™ 10L Round Polypropylene (PP) Carboy, 83B (83mm) VersaCap Closure w/ Spigot
GREENER_CHOICE
Catalog No. NC1818183
Encompass
Foxx Life Sciences offers a premium line of high quality, Round Polypropylene (PP) laboratory Carboys. All round carboys feature Foxx's patented VersaCap technology. VersaCap offers large handles with finger grips that allow for easy tightening and opening. Our versatile leak proof carboys with strong easy grip handles, aim to take full advantage of storage efficiency at minimal effort. Round carboys are available in 10 Liter and 20 Liter options, wih or without a leak-proof spigot. Each carboy also comes with an 83B Polypropylene (PP) VersaCap closure. 20 Liter Carboy comes with Graduation marks. Foxx laboratory Polypropylene (PP) carboys are made from a USP Class VI, FDA Grade, autoclavable and Heavy Metal Free material. The recommended autoclaving cycle is 15 PSI for 20 minutes at 121C (250F).
Catalog No. NC1818183 Supplier Foxx Life Sciences Supplier No. 155B817FLS
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Foxx Life Sciences offers a premium line of high quality, Round Polypropylene (PP) laboratory Carboys. All round carboys feature Foxx's patented VersaCap technology. VersaCap offers large handles with finger grips that allow for easy tightening and opening. Our versatile leak proof carboys with strong easy grip handles, aim to take full advantage of storage efficiency at minimal effort. Round carboys are available in 10 Liter and 20 Liter options, wih or without a leak-proof spigot. Each carboy also comes with an 83B Polypropylene (PP) VersaCap closure. 20 Liter Carboy comes with Graduation marks. Foxx laboratory Polypropylene (PP) carboys are made from a USP Class VI, FDA Grade, autoclavable and Heavy Metal Free material. The recommended autoclaving cycle is 15 PSI for 20 minutes at 121C (250F).