Learn More
Foxx Life Sciences EZBio® 10L (2.5 GAL) Gamma Sterilized Double Bagged Light Blocking Amber HDPE Carboy with 83mm (83B) VersaCap® Closure
GREENER_CHOICE
Catalog No. 501720660
Encompass
Foxx Life Sciences EZBio® Gamma Sterile Amber HDPE Carboys provide users with a highly versatile container that maximizes storage efficiency and ease of use. Carboys are made entirely with USP Class VI components and Gamma Irradiated in the United States. The distinctive rectangular shape saves valuable bench space. Large handles on top of the carboy and molded grips in the base provide a full, secure grip to make lifting, handling, and pouring easy. The optional wide neck allows users to reach inside the carboy for easier cleaning. Carboy material and volume are clearly imprinted on the outside of the bottle, along with large, easy to read metric molded graduation marks which are certified to 5% accuracy.Amber HDPE carboys are made from a USP Class VI, FDA Grade materials. EZgrip® ergonomic shape maximizes bench space.EZfil® wide-mouth design for easy filling and cleaning.VersaCap® offers a wide variety of top connection options. 13L Max Capacity.
Catalog No. 501720660 Supplier Foxx Life Sciences Supplier No. 51K5G51FLS
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Foxx Life Sciences EZBio® Gamma Sterile Amber HDPE Carboys provide users with a highly versatile container that maximizes storage efficiency and ease of use. Carboys are made entirely with USP Class VI components and Gamma Irradiated in the United States. The distinctive rectangular shape saves valuable bench space. Large handles on top of the carboy and molded grips in the base provide a full, secure grip to make lifting, handling, and pouring easy. The optional wide neck allows users to reach inside the carboy for easier cleaning. Carboy material and volume are clearly imprinted on the outside of the bottle, along with large, easy to read metric molded graduation marks which are certified to 5% accuracy.Amber HDPE carboys are made from a USP Class VI, FDA Grade materials. EZgrip® ergonomic shape maximizes bench space.EZfil® wide-mouth design for easy filling and cleaning.VersaCap® offers a wide variety of top connection options. 13L Max Capacity.