missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carboxymethoxylamine Hemihydrochloride 98.0+%, TCI America™
Supplier: TCI America C0205500G
Specifications
| Carboxymethoxylamine Hemihydrochloride | |
| White | |
| MFCD00012955 | |
| o-carboxymethyl hydroxylamine hemihydrochloride, carboxymethoxylamine hemihydrochloride, aminooxyacetic acid hemihydrochloride, 2-aminooxy acetic acid hydrochloride 2:1, unii-5g5qi2gn7a, acetic acid, aminooxy-, hydrochloride 2:1, aminooxy acetic acid hemihydrochloride, 5g5qi2gn7a, carboxymethoxyamine hemihydrochloride | |
| [Cl-].NOCC(O)=O.[NH3+]OCC(O)=O | |
| 218.59 | |
| 109.30 | |
| Crystalline Powder |
| 2921-14-4 | |
| C4H11ClN2O6 | |
| 500 g | |
| AXIKLJPWEGNZIG-UHFFFAOYSA-N | |
| 2-(aminooxy)acetic acid 2-(azaniumyloxy)acetic acid chloride | |
| 2723609 | |
| ≥98.0% (T) |
Chemical Identifiers
| 2921-14-4 | |
| 218.59 | |
| AXIKLJPWEGNZIG-UHFFFAOYSA-N | |
| 2723609 | |
| [Cl-].NOCC(O)=O.[NH3+]OCC(O)=O |
| C4H11ClN2O6 | |
| MFCD00012955 | |
| o-carboxymethyl hydroxylamine hemihydrochloride, carboxymethoxylamine hemihydrochloride, aminooxyacetic acid hemihydrochloride, 2-aminooxy acetic acid hydrochloride 2:1, unii-5g5qi2gn7a, acetic acid, aminooxy-, hydrochloride 2:1, aminooxy acetic acid hemihydrochloride, 5g5qi2gn7a, carboxymethoxyamine hemihydrochloride | |
| 2-(aminooxy)acetic acid 2-(azaniumyloxy)acetic acid chloride |
Safety and Handling
EINECSNumber : (2)-3700
RTECSNumber : AF3150000
TSCA : Yes