missing translation for 'onlineSavingsMsg'
Learn More

Carboxymethoxylamine Hemihydrochloride 98.0+%, TCI America™

Catalog No. C020525G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C020525G 25 g
C0205500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C020525G Supplier TCI America Supplier No. C020525G

Chemical Identifiers

CAS 2921-14-4
Molecular Formula C4H11ClN2O6
Molecular Weight (g/mol) 218.59
MDL Number MFCD00012955
InChI Key AXIKLJPWEGNZIG-UHFFFAOYSA-N
Synonym o-carboxymethyl hydroxylamine hemihydrochloride, carboxymethoxylamine hemihydrochloride, aminooxyacetic acid hemihydrochloride, 2-aminooxy acetic acid hydrochloride 2:1, unii-5g5qi2gn7a, acetic acid, aminooxy-, hydrochloride 2:1, aminooxy acetic acid hemihydrochloride, 5g5qi2gn7a, carboxymethoxyamine hemihydrochloride
PubChem CID 2723609
IUPAC Name 2-(aminooxy)acetic acid 2-(azaniumyloxy)acetic acid chloride
SMILES [Cl-].NOCC(O)=O.[NH3+]OCC(O)=O

Specifications

Color White
Quantity 25 g
Formula Weight 109.30
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Carboxymethoxylamine Hemihydrochloride
EINECSNumber (2)-3700
RTECSNumber AF3150000
TSCA Yes