missing translation for 'onlineSavingsMsg'
Learn More

Carboxy-PTIO potassium salt, 98+%

Catalog No. AAJ61316MA
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ61316MA 10 mg
AAJ61316MF 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ61316MA Supplier Thermo Scientific Chemicals Supplier No. J61316MA

Stable, water-soluble free radical that reacts with nitric oxide (NO) to inhibit NO synthase | CAS: 148819-94-7 | C14H17KN2O4 | 316.398 g/mol

Stable, water-soluble free radical that reacts with nitric oxide (NO) to inhibit NO synthase

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 148819-94-7
Molecular Formula C14H17KN2O4
Molecular Weight (g/mol) 316.398
MDL Number MFCD00216153
InChI Key CUYIQXAMAUOMKG-UHFFFAOYSA-N
Synonym potassium ion 2-4-carboxylatophenyl-3-hydroxy-4,4,5,5-tetramethylimidazol-1-ium-1-olate, 2-4-potassiooxycarbonyl phenyl-4,4,5,5-tetramethyl-2-imidazoline 3-oxide-1-yl oxy radical, potassium 2-4-carboxylatophenyl-3-hydroxy-4,4,5,5-tetramethylimidazol-1-ium-1-olate, potassium 2-4-carboxylatophenyl-4,4,5,5-tetramethyl-3-oxidoimidazol-3-ium-1-yloxidanyl
PubChem CID 54730022
IUPAC Name potassium;4-(4,4,5,5-tetramethyl-1,3-dioxidoimidazol-1-ium-2-yl)benzoic acid
SMILES CC1(C([N+](=C(N1[O-])C2=CC=C(C=C2)C(=O)O)[O-])(C)C)C.[K+]

Specifications

Color Blue
Quantity 10 mg
Solubility Information Soluble in water; Also soluble in ethanol,methanol,DMSO or DMF.
Formula Weight 315.38
Percent Purity ≥98%
Physical Form Solid
Chemical Name or Material Carboxy-PTIO potassium salt
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only