missing translation for 'onlineSavingsMsg'
Learn More

Carbowax™ PEG 8000 (Powder), Fisher Chemical™

Catalog No. p-82261
Change view
Click to view available options
Quantity:
500 g
3 kg
Packaging:
Poly Bottle
Poly Pail
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
P156500 500 g Poly Bottle
P1563 3 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P156500 Supplier Thermo Fisher Scientific Supplier No. P156 500
Only null left

Polyethylene Glycol

Chemical Identifiers

CAS 25322-68-3
Molecular Formula (C2H4O)n
Molecular Weight (g/mol) 62.07
MDL Number MFCD01779601
InChI Key LYCAIKOWRPUZTN-UHFFFAOYSA-N
Synonym Polyethylene Glycol, Macrogol
IUPAC Name ethane-1,2-diol
SMILES [H]OCCO

Specifications

Color White
pH 5.0 to 7.0
Physical Form Solid
Boiling Point 200°C
Melting Point 55°C to 60°C
Density 1.1g/mL
Quantity 500 g
Formula Weight 7000 to 9000
Flash Point 270°C
Identification Pass Test
Packaging Poly Bottle
Chemical Name or Material Carbowax™ PEG 8000
Show More Show Less
Hazard Statement CAUTION!
Precautionary Statement Emergency Overview
May cause skin, eye, and respiratory tract irritation. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. Obtain medical attention. .

NFPA

Health:1
Flammability:1
Instability:0