missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Carbonate-buffered saline (5X), pH 9.5
$120.29 - $120.29
Specifications
| Concentration | 5X |
|---|---|
| pH Range | 9.5 |
| Format | Liquid |
| Storage Requirements | Ambient temperatures |
| Type | Carbonate-buffered saline |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAJ63928AP
|
Thermo Scientific™
J63928AP |
500 mL |
Each for $120.29
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
| 5X | |
| Liquid | |
| Carbonate-buffered saline |
| 9.5 | |
| Ambient temperatures |