missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Carbonate, 0.5M buffer soln., pH 9.0

Catalog No. AAJ63899AK
Change view
Click to view available options
Quantity:
250 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ63899AK 250 mL
AAJ63899AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ63899AK Supplier Thermo Scientific Chemicals Supplier No. J63899AK

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Concentration 0.5 M buffer soln.
pH Range 9.0
Quantity 250 mL
Type Carbonate buffer soln.

RUO – Research Use Only