missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Carbonate, 0.2M buffer soln., pH 9.4, low endotoxin

Catalog No. AAJ67013AE
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ67013AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ67013AE Supplier Thermo Scientific™ Supplier No. J67013AE

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Color Colorless
Concentration 0.2 M buffer soln.
pH Range 9.4
Format Liquid
Quantity 100 mL
Storage Requirements Ambient temperatures
Type Carbonate