missing translation for 'onlineSavingsMsg'
Learn More

Carbon felt, 6.35mm (0.25in) thick, 99.0%, Thermo Scientific Chemicals

Catalog No. AA43200RR
Change view
Click to view available options
Quantity:
10 x 10 cm
30 x 30 cm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA43200RR 10 x 10 cm
AA43200RF 30 x 30 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA43200RR Supplier Thermo Scientific Chemicals Supplier No. 043200RR

Typical thermal conductivity of 1.75 BTU in/hr/ft²/°F

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1333-86-4
Molecular Formula C
Molecular Weight (g/mol) 12.01
MDL Number MFCD00133992
InChI Key OKTJSMMVPCPJKN-UHFFFAOYSA-N

Specifications

Density 2.26g/cm3
Assay Percent Range 99.0%
Quantity 10 x 10 cm
Chemical Name or Material Carbon
EINECSNumber 231-955-3
TSCA U
Recommended Storage Ambient temperatures

RUO – Research Use Only