missing translation for 'onlineSavingsMsg'
Learn More

Carbon black, Super P∣r Conductive, 99+% (metals basis)

Catalog No. AAH3025314
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3025314 25 g
AAH3025322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3025314 Supplier Thermo Scientific Chemicals Supplier No. H3025314

Carbon black, Super P, >-99%, CAS Number-1333-86-4, 25g, (metals basis), Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S., 215-609-9, 12.01, H351, GHS H Statement H228-H351 Flammable solid. Suspected of causing cancer. | CAS: 1333-86-4 | C | 12.01 g/mol

Carbon black Super-P (TIMCAL) was used as conductive agent. Super P furnace black the best conductive additive. Carbon black (conducting material, super P black) was added with binder in the composite electrode to compensate the low electrical conductivity of PPy and PPyDVB in miniemulsion polymerization. The hybrid Super P-SACNT conductive network manifests itself as a promising strategy to improve the battery performances with a minimum amount of conductive fillers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Carbon black Super-P (TIMCAL) was used as conductive agent. Super P furnace black the best conductive additive. Carbon black (conducting material, super P black) was added with binder in the composite electrode to compensate the low electrical conductivity of PPy and PPyDVB in miniemulsion polymerization. The hybrid Super P-SACNT conductive network manifests itself as a promising strategy to improve the battery performances with a minimum amount of conductive fillers.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 1333-86-4
Molecular Formula C
Molecular Weight (g/mol) 12.01
MDL Number MFCD00133992
InChI Key OKTJSMMVPCPJKN-UHFFFAOYSA-N

Specifications

Odor Odorless
Quantity 25 g
UN Number UN1325
Formula Weight 12.01
Percent Purity ≥99%
Physical Form Powder
Assay (metals basis)
Chemical Name or Material Carbon black, Super P™, Conductive
Hazard Category H351
Hazard Statement GHS H Statement
H228-H351
Flammable solid.
Suspected of causing cancer.
Precautionary Statement P201-P202-P281-P308+P313-P501c
DOTInformation Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.
EINECSNumber 215-609-9
RTECSNumber FF5800000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only