missing translation for 'onlineSavingsMsg'
Learn More

Carbon black, acetylene, 100% compressed

Catalog No. p-4407517
Change view
Click to view available options
Quantity:
250 g
2 x 500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4552730 250 g
AA4552795 2 x 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4552730 Supplier Thermo Scientific Chemicals Supplier No. 04552730
Only null left

CAS: 1333-86-4 | C | 12.01 g/mol

It is used in optical computing, optical memories, optical signal processing and controlling applications respects, C60 and its derivatives can be widely used in photovoltaic applications, magnetic resonance imaging, anti-HIV drugs, anti-cancer drugs, chemotherapy drugs, cosmetic additives, research and other areas.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Chemical Identifiers

CAS 1333-86-4
Molecular Formula C
Molecular Weight (g/mol) 12.01
MDL Number MFCD00133992
InChI Key OKTJSMMVPCPJKN-UHFFFAOYSA-N

Specifications

Color Black
Density 1.8 to 2.1g/cm3 at 20°C
Assay Percent Range >99.9%
Quantity 250 g
Physical Form Powder
Chemical Name or Material Carbon black
Hazard Category H351
Precautionary Statement P201-P202-P281-P308+P313-P501c
DOTInformation DOT Class: 4.1, Packaging Group: III
EINECSNumber 215-609-9
RTECSNumber FF5800000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only