missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Carbon Adsorbent Sampler Kit matrix CMS/SGPC Kit I

Catalog No. 111005910
Change view
Click to view available options
8 x 5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
111005910 8 x 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111005910 Supplier MilliporeSigma™ Supelco™ Supplier No. 13028U
Only null left

A cost-effective way to evaluate several of Supelco's specialty carbon adsorbents.

Choosing the right adsorbent or combination of adsorbents can be difficult: Must choose one or more to retain specific analyte(s). May be equally important to release analyte(s) during desorption.

Specifications

Packaging Bottle
Quantity 8 x 5 g