Learn More
Carbon, Activated (Powder/USP), Fisher Chemical™
Fine black powder, free from gritty matter
Supplier: Thermo Fisher Scientific FLC272212
Specifications
| Carbon, Activated | |
| 3652°C | |
| 4.0% max. | |
| Poly Pail | |
| MFCD00133992 | |
| graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve | |
| Meets Requirements | |
| [C] | |
| 12.011 | |
| CHEBI:27594 | |
| USP |
| 7440-44-0 | |
| Black | |
| 15.0% max. | |
| C | |
| 2.5 kg | |
| 3.5% max. Acid Soluble Substances | |
| OKTJSMMVPCPJKN-UHFFFAOYSA-N | |
| carbon | |
| 5462310 | |
| 1mmHg at 3586°C | |
| Solid |
Chemical Identifiers
| 7440-44-0 | |
| 12.011 | |
| OKTJSMMVPCPJKN-UHFFFAOYSA-N | |
| 5462310 | |
| carbon |
| C | |
| MFCD00133992 | |
| graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve | |
| CHEBI:27594 | |
| [C] |
Safety and Handling
CAUTION!
Emergency Overview
May cause skin and eye irritation. Avoid contact with skin and eyes. Use personal protective equipment. Look out! DATATYPE 710. Avoid breathing dust/fume/gas/mist/vapours/spray. Rinse with water. . Obtain medical attention. Remove from exposure, lie down. If breathing is difficult, give oxygen. If not breathing, give artificial respiration. If not breathing, give artificial respiration. Obtain medical attention.
NFPA
Health:1
Flammability:1
Instability:0