missing translation for 'onlineSavingsMsg'
Learn More

Carbon, Activated (Powder/USP), Fisher Chemical™

Catalog No. C272500
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Glass Bottle
Poly Pail
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
C272500 500 g Glass Bottle
C272212 2.5 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C272500 Supplier Thermo Fisher Scientific Supplier No. FLC272 500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Fine black powder, free from gritty matter

Chemical Identifiers

CAS 7440-44-0
Molecular Formula C
Molecular Weight (g/mol) 12.011
MDL Number MFCD00133992
InChI Key OKTJSMMVPCPJKN-UHFFFAOYSA-N
Synonym graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve
PubChem CID 5462310
ChEBI CHEBI:27594
IUPAC Name carbon
SMILES [C]

Specifications

Melting Point 3652°C
Color Black
Density 1.8g/mL
Ignition Residue 4.0% max.
Loss on Drying 15.0% max.
Packaging Glass Bottle
Quantity 500 g
Solubility Information 3.5% max. Acid Soluble Substances
Residual Solvents Meets Requirements
Vapor Pressure 1mmHg at 3586°C
Grade USP
Physical Form Solid
Chemical Name or Material Carbon, Activated
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Statement CAUTION!
Precautionary Statement Emergency Overview
May cause skin and eye irritation. Avoid contact with skin and eyes. Use personal protective equipment. Look out! DATATYPE 710. Avoid breathing dust/fume/gas/mist/vapours/spray. Rinse with water. . Obtain medical attention. Remove from exposure, lie down. If breathing is difficult, give oxygen. If not breathing, give artificial respiration. If not breathing, give artificial respiration. Obtain medical attention.

NFPA

Health:1
Flammability:1
Instability:0