missing translation for 'onlineSavingsMsg'
Learn More

Carbon, activated, Norit ROW 0.8mm pellets, steam activated

CAS: 7440-44-0 | C | 12.011 g/mol

Supplier:  Thermo Scientific Chemicals L163340P

 View more versions of this product

Catalog No. AAL163340P


Only null left
Explore available promotions

Description

Description

Carbon, activated, Norit ROW 0.8mm pellets, steam activated is suitable for decolorization, deodorization and purification applications. It is mainly used for gas phase applications such as automotive emission control due to their low pressure drop, high mechanical strength and low dust content. It has high removal efficiency for removing degradation byproducts for drinking water. It also removes foaming causing impurities for the purification of amine and glycol process streams.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Carbon, activated, Norit ROW 0.8mm pellets, steam activated is suitable for decolorization, deodorization and purification applications. It is mainly used for gas phase applications such as automotive emission control due to their low pressure drop, high mechanical strength and low dust content. It has high removal efficiency for removing degradation byproducts for drinking water. It also removes foaming causing impurities for the purification of amine and glycol process streams.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with acids and oxidizing agents.
Specifications

Specifications

Carbon
7440-44-0
MFCD00133992
graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve
OKTJSMMVPCPJKN-UHFFFAOYSA-N
carbon
5462310
activated, Norit ROW 0.8mm pellets, steam activated
C
2000 g
Insoluble in water.
[C]
12.011
CHEBI:27594

Chemical Identifiers

7440-44-0
12.011
OKTJSMMVPCPJKN-UHFFFAOYSA-N
5462310
carbon
C
MFCD00133992
graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve
CHEBI:27594
[C]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

H500

EINECSNumber : 231-153-3

RTECSNumber : FF5250100

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only