missing translation for 'onlineSavingsMsg'
Learn More

Carbon, activated, Norit ROW 0.8mm pellets, steam activated

Catalog No. AAL1633436
Change view
Click to view available options
Quantity:
100 g
500 g
2000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1633436 500 g
AAL1633422 100 g
AAL163340P 2000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1633436 Supplier Thermo Scientific Chemicals Supplier No. L1633436

CAS: 7440-44-0 | C | 12.011 g/mol

Carbon, activated, Norit ROW 0.8mm pellets, steam activated is suitable for decolorization, deodorization and purification applications. It is mainly used for gas phase applications such as automotive emission control due to their low pressure drop, high mechanical strength and low dust content. It has high removal efficiency for removing degradation byproducts for drinking water. It also removes foaming causing impurities for the purification of amine and glycol process streams.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Carbon, activated, Norit ROW 0.8mm pellets, steam activated is suitable for decolorization, deodorization and purification applications. It is mainly used for gas phase applications such as automotive emission control due to their low pressure drop, high mechanical strength and low dust content. It has high removal efficiency for removing degradation byproducts for drinking water. It also removes foaming causing impurities for the purification of amine and glycol process streams.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with acids and oxidizing agents.

Chemical Identifiers

CAS 7440-44-0
Molecular Formula C
Molecular Weight (g/mol) 12.011
MDL Number MFCD00133992
InChI Key OKTJSMMVPCPJKN-UHFFFAOYSA-N
Synonym graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve
PubChem CID 5462310
ChEBI CHEBI:27594
IUPAC Name carbon
SMILES [C]

Specifications

Quantity 500 g
Solubility Information Insoluble in water.
Chemical Name or Material Carbon, activated, Norit ROW 0.8mm pellets, steam activated
Hazard Category H500
EINECSNumber 231-153-3
RTECSNumber FF5250100
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only