missing translation for 'onlineSavingsMsg'
Learn More

Carbon, activated, -20+40 mesh

Catalog No. p-4407519
Change view
Click to view available options
Quantity:
100 g
500 g
2 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4547822 100 g
AA4547836 500 g
AA45478A3 2 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4547822 Supplier Thermo Scientific Chemicals Supplier No. 04547822
Only null left

CAS: 7440-44-0 | C | 12.011 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-44-0
Molecular Formula C
Molecular Weight (g/mol) 12.011
MDL Number MFCD00133992
InChI Key OKTJSMMVPCPJKN-UHFFFAOYSA-N
Synonym graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve
PubChem CID 5462310
ChEBI CHEBI:27594
IUPAC Name carbon
SMILES [C]

Specifications

Color Black
Mesh Size -20+40 Mesh
Quantity 100 g
Physical Form Powder
Chemical Name or Material Carbon
Hazard Category H500
EINECSNumber 231-153-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only