missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbomer 941, NF, Spectrum™ Chemical
Description
Spectrum™ Chemical Carbomer 941, NF is an expanded molecule that will increase continually when more alkyne bonds are presented. It is because of this that carbomers are also called carbon-molecules. The NF grade indicates it is graded suitable for personal care, cosmetic and pharmaceutical applications and were manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
| CAS Max % | 100% |
| Quantity | 50 lb. |
| Grade | NF |
| Loss on Drying | 2% |
| Packaging | Fiber Drum |