missing translation for 'onlineSavingsMsg'
Learn More

Carbohydrazide 98.0+%, TCI America™

Catalog No. p-7124618
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C080325G 25 g
C0803500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C080325G Supplier TCI America Supplier No. C080325G
Only null left

Chemical Identifiers

CAS 497-18-7
Molecular Formula CH6N4O
Molecular Weight (g/mol) 90.09
MDL Number MFCD00007591
InChI Key XEVRDFDBXJMZFG-UHFFFAOYSA-N
Synonym carbohydrazide, carbonic dihydrazide, carbazide, carbonohydrazide, carbodihydrazide, hydrazine, carbonyldi, hydrazine, carbonylbis, carbazic acid, hydrazide, semicarbazide, 4-amino, carbonic acid, dihydrazide
PubChem CID 73948
ChEBI CHEBI:61308
IUPAC Name 1,3-diaminourea
SMILES NNC(=O)NN

Specifications

Melting Point 155°C
Color White
Quantity 25 g
UN Number 3077
Formula Weight 90.09
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Carbohydrazide
EINECSNumber (2)-3345
RTECSNumber FF2625000
TSCA Yes
Recommended Storage Refrigerator