missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Carbohydrate Sulfotransferase 3/CHST3 Recombinant Protein Antigen

Catalog No. NBP57128PEP
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP57128PEP 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP57128PEP Supplier R&D Systems™ Supplier No. NBP257128PEP

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A Novus product, part of R&D Systems. Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition

A recombinant protein antigen with a N-terminal His6-ABP tag corresponding to human CHST3. The Carbohydrate Sulfotransferase 3/CHST3 Recombinant Protein Antigen has been validated for the following applications: Antibody Competition.

Specifications

Gene ID (Entrez) 9469
Product Line Novus
Purity >80% by SDS-PAGE and Coomassie blue staining
Common Name C6ST-1, C6ST1HSD, C6STEC 2.8.2.17, carbohydrate (chondroitin 6) sulfotransferase 3, carbohydrate sulfotransferase 3, Chondroitin 6-O-sulfotransferase 1, Chondroitin 6-sulfotransferase, EC 2.8.2, Galactose/N-acetylglucosamine/N-acetylglucosamine 6-O-sulfotransferase 0, GST-0
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Gene Symbol CHST3
Molecular Weight (g/mol) 23 kDa
Quantity 100 μL
Research Category Signal Transduction
Source E. coli
Specific Reactivity This recombinant antigen is only intended to be used as a blocking agent to confirm antibody specificity with the corresponding antibody, catalog number NBP2-54239. It is purified by IMAC chromatography, and the expected concentration is greater than 0.5 mg/ml
Show More Show Less