missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbidopa Monohydrate 98.0+%, TCI America™
$46.60 - $136.08
Chemical Identifiers
| CAS | 38821-49-7 |
|---|---|
| Molecular Formula | C10H16N2O5 |
| Molecular Weight (g/mol) | 244.247 |
| MDL Number | MFCD00889211 |
| InChI Key | QTAOMKOIBXZKND-PPHPATTJSA-N |
| Synonym | 3-3,4-dihydroxyphenyl-2-hydrazinyl-2-methylpropanoic acid hydrate, +-2-3,4-dihydroxybenzyl-2-hydrazinopropionic acid monohydrate, 3-3,4-bis oxidanyl phenyl-2-diazanyl-2-methyl-propanoic acid hydrate |
| PubChem CID | 38101 |
| ChEBI | CHEBI:3395 |
| IUPAC Name | (2S)-3-(3,4-dihydroxyphenyl)-2-hydrazinyl-2-methylpropanoic acid;hydrate |
| SMILES | CC(CC1=CC(=C(C=C1)O)O)(C(=O)O)NN.O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
C2450100MG
|
TCI America
C2450100MG |
100 mg |
Each for $46.60
|
|
|||||
|
C24501G
|
TCI America
C24501G |
1 g |
Each for $136.08
|
|
|||||
Chemical Identifiers
| 38821-49-7 | |
| 244.247 | |
| QTAOMKOIBXZKND-PPHPATTJSA-N | |
| 38101 | |
| (2S)-3-(3,4-dihydroxyphenyl)-2-hydrazinyl-2-methylpropanoic acid;hydrate |
| C10H16N2O5 | |
| MFCD00889211 | |
| 3-3,4-dihydroxyphenyl-2-hydrazinyl-2-methylpropanoic acid hydrate, +-2-3,4-dihydroxybenzyl-2-hydrazinopropionic acid monohydrate, 3-3,4-bis oxidanyl phenyl-2-diazanyl-2-methyl-propanoic acid hydrate | |
| CHEBI:3395 | |
| CC(CC1=CC(=C(C=C1)O)O)(C(=O)O)NN.O |
Specifications
| 38821-49-7 | |
| C10H16N2O5 | |
| 100 mg | |
| QTAOMKOIBXZKND-PPHPATTJSA-N | |
| (2S)-3-(3,4-dihydroxyphenyl)-2-hydrazinyl-2-methylpropanoic acid;hydrate | |
| 38101 | |
| 226.23 | |
| Crystalline Powder |
| White-Yellow | |
| MFCD00889211 | |
| 3-3,4-dihydroxyphenyl-2-hydrazinyl-2-methylpropanoic acid hydrate, +-2-3,4-dihydroxybenzyl-2-hydrazinopropionic acid monohydrate, 3-3,4-bis oxidanyl phenyl-2-diazanyl-2-methyl-propanoic acid hydrate | |
| CC(CC1=CC(=C(C=C1)O)O)(C(=O)O)NN.O | |
| 244.247 | |
| CHEBI:3395 | |
| ≥98.0% (HPLC,T) | |
| Carbidopa Monohydrate |
Safety and Handling
RTECSNumber : MW5300000
TSCA : No