missing translation for 'onlineSavingsMsg'
Learn More

Carbidopa Monohydrate 98.0+%, TCI America™

Catalog No. C24501G
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C24501G 1 g
C2450100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C24501G Supplier TCI America Supplier No. C24501G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 38821-49-7
Molecular Formula C10H16N2O5
Molecular Weight (g/mol) 244.247
MDL Number MFCD00889211
InChI Key QTAOMKOIBXZKND-PPHPATTJSA-N
Synonym 3-3,4-dihydroxyphenyl-2-hydrazinyl-2-methylpropanoic acid hydrate, +-2-3,4-dihydroxybenzyl-2-hydrazinopropionic acid monohydrate, 3-3,4-bis oxidanyl phenyl-2-diazanyl-2-methyl-propanoic acid hydrate
PubChem CID 38101
ChEBI CHEBI:3395
IUPAC Name (2S)-3-(3,4-dihydroxyphenyl)-2-hydrazinyl-2-methylpropanoic acid;hydrate
SMILES CC(CC1=CC(=C(C=C1)O)O)(C(=O)O)NN.O

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 226.23
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Carbidopa Monohydrate
RTECSNumber MW5300000
TSCA No