missing translation for 'onlineSavingsMsg'
Learn More

Carbenoxolone disodium salt, 97+%

Catalog No. AAJ6371406
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6371406 5 g
AAJ6371403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6371406 Supplier Thermo Scientific Chemicals Supplier No. J6371406
Only null left

CAS: 7421-40-1 | C34H48Na2O7 | 614.73 g/mol

Carbenoxolone is anti-inflammatory. It is often used as a gap junction inhibitor in neuroscience research. Carbenoxolone is also an inhibitor of the steroid dehydrogenase enzymes 11 β-hydroxysteroid dehydrogenase (human) and 3α, 20 β-hydroxysteroid dehydrogenase (bacterial). It is used for the treatment of digestive tract ulcers, especially in the stomach. Used in the treatment of peptic, mainly gastric, ulcers. It is often used as a gap junction inhibitor in neuroscience research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water to 100 mM.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from heat.

Chemical Identifiers

CAS 7421-40-1
Molecular Formula C34H48Na2O7
Molecular Weight (g/mol) 614.73
MDL Number MFCD00079043
InChI Key BQENDLAVTKRQMS-SBBGFIFASA-L
Synonym carbenoxolone disodium salt, disodium 3beta-3-3-carboxylatopropanoyl oxy-11-oxoolean-12-en-30-oate, disodium 4as,6as,6br,10s,12as,12br,14br-10-3-carboxylatopropanoyl oxy-2,4a,6a,6b,9,9,12a-heptamethyl-13-oxo-3,4,5,6,7,8,8a,10,11,12,12b,14b-dodecahydro-1h-picene-2-carboxylate
PubChem CID 133687779
IUPAC Name disodium;(2R,4aR,6aS,6aR,6bS,8aS,10R,12aR,14bS)-10-(3-carboxylatopropanoyloxy)-2,4a,6a,6b,9,9,12a-heptamethyl-13-oxo-3,4,5,6,6a,7,8,8a,10,11,12,14b-dodecahydro-1H-picene-2-carboxylate
SMILES [Na+].[Na+].[H][C@@]12C[C@](C)(CC[C@]1(C)CC[C@]1(C)C2=CC(=O)[C@]2([H])[C@@]3(C)CC[C@H](OC(=O)CCC([O-])=O)C(C)(C)[C@]3([H])CC[C@@]12C)C([O-])=O

Specifications

Color White to Cream
Quantity 5 g
Merck Index 14,1793
Solubility Information Soluble in water to 100 mM.
Formula Weight 614.7
Percent Purity ≥97%
Physical Form Solid
Chemical Name or Material Carbenoxolone disodium salt
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 231-044-0
RTECSNumber RK0250000
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only