missing translation for 'onlineSavingsMsg'
Learn More

Carbenicillin disodium salt

Catalog No. AAJ6194914
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6194914 25 g
AAJ6194903 1 g
AAJ6194906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6194914 Supplier Thermo Scientific Chemicals Supplier No. J6194914

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Carbenicillin disodium salt, CAS # 4800-94-6, is the disodium salt form of carbenicillin, a semisynthetic compound with broad-spectrum antibacterial and β-lactamase-resistant activities. | CAS: 4800-94-6 | C17H16N2Na2O6S | 422.36 g/mol

Carbenicillin disodium salt is a water-soluble antibiotic which is effective against gram-negative bacteria.Carbenicillin disodium salt is widely utilized as an antibiotic which is used to control bacterial cell-wall synthesis by inactivating transpeptidases on the inner surface of the bacterial cell membrane. It has been involved in the study of the role of penicillin-sensitive transpeptidases in cell wall biosynthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Carbenicillin disodium salt is a semisynthetic compound with antibacterial and β-lactamase-resistant activities and is effective against gram-negative bacteria
  • In in vitro studies, carbenicillin disodium salt can totally suppress the proliferation of Agrobacterium

Application

  • Due to its antibacterial activity, this compound can be used to control bacterial cell wall synthesis by inactivating transpeptidases on the inner surface of the bacterial cell membrane. This can lead to incomplete bacterial cell wall synthesis to eventually cause cell lysis
  • It has also been used in studies to evaluate the role of penicillin-sensitive transpeptidases in cell wall biosynthesis

Chemical Identifiers

CAS 4800-94-6
Molecular Formula C17H16N2Na2O6S
Molecular Weight (g/mol) 422.36
MDL Number MFCD00077683
InChI Key RTYJTGSCYUUYAL-YCAHSCEMSA-L
PubChem CID 20933
ChEBI CHEBI:34609
SMILES [Na+].[Na+].CC1(C)S[C@@H]2[C@H](NC(=O)C(C([O-])=O)C3=CC=CC=C3)C(=O)N2[C@H]1C([O-])=O

Specifications

Type Carbenicillin Disodium Salt
Physical Form Powder
Color White
Quantity 25 g
Beilstein 5722128
Merck Index 14,1792
Solubility Information Soluble in water and alcohol.
IUPAC Name disodium (2S,5R,6R)-6-(2-carboxylato-2-phenylacetamido)-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
Formula Weight 422.36
Chemical Name or Material Carbenicillin disodium salt
Hazard Category H317-H334-H335
Hazard Statement GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
Precautionary Statement P261-P271-P272-P280g-P285-P302+P352-P304+P340-P333+P313-P342+P311-P363-P501c
EINECSNumber 225-360-8
RTECSNumber ON9105000
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only