missing translation for 'onlineSavingsMsg'
Learn More

Carbenicillin (Disodium Salt), Fisher BioReagents

Catalog No. p-25005
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BP2648250 250 mg Amber Glass
BP26481 1 g Amber Glass
BP26485 5 g Amber Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. BP2648250 Supplier Fisher BioReagents Supplier No. BP2648250
Used for the selection of ampicillin-resistant transformed cells and for the study of penicillin-sensitive transpeptidases in cell wall biosynthesis. Antimicrobial spectrum: Gram-positive and Gram-negative bacteria, Pseudomonas.

Chemical Identifiers

CAS 4800-94-6
Molecular Formula C17H16N2Na2O6S
Molecular Weight (g/mol) 422.36
MDL Number MFCD00077683
InChI Key RTYJTGSCYUUYAL-YCAHSCEMSA-L
Synonym α-Carboxybenzylpenicillin
PubChem CID 20933
ChEBI CHEBI:34609
IUPAC Name disodium (2S,5R,6R)-6-(2-carboxylato-2-phenylacetamido)-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
SMILES [Na+].[Na+].CC1(C)S[C@@H]2[C@H](NC(=O)C(C([O-])=O)C3=CC=CC=C3)C(=O)N2[C@H]1C([O-])=O

Specifications

Type Carbenicillin
Physical Form Solid
Melting Point 200°C
Packaging Amber Glass
Potency 770μg/mg min.
Water 6% Max.
Color White
Quantity 250 mg
Assay Percent Range ≥92 %
Boiling Point 738°C
Formula Weight 422.4
Percent Purity 92% min.
Chemical Name or Material Carbenicillin, Disodium Salt
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Statement WARNING!
Precautionary Statement Emergency Overview
May cause allergic respiratory and skin reaction. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required. Do not induce vomiting. Do not induce vomiting. Call a physician or Poison Control Center immediately.

NFPA

Health:2
Flammability:0
Instability:0
Recommended Storage 0° to 5°C