missing translation for 'onlineSavingsMsg'
Learn More

Carbazole 97.0+%, TCI America™

Catalog No. c003225g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
25 g
500 g
Catalog No. Quantity
C003225G 25 g
C0032100G 100 g
C0032500G 500 g
3 options

Catalog No. C003225G

Supplier: TCI America C003225G

Only null left

Chemical Identifiers

CAS 86-74-8
Molecular Formula C12H9N
Molecular Weight (g/mol) 167.21
MDL Number MFCD00004960
InChI Key UJOBWOGCFQCDNV-UHFFFAOYSA-N
Synonym carbazole, dibenzopyrrole, diphenylenimine, 9-azafluorene, diphenylenimide, diphenyleneimine, dibenzo b,d pyrrole, usaf ek-600, unii-0p2197hhhn
PubChem CID 6854
ChEBI CHEBI:27543
IUPAC Name 9H-carbazole
SMILES N1C2=C(C=CC=C2)C2=C1C=CC=C2

Specifications

Melting Point 245°C
Color White-Yellow
Boiling Point 355°C
Quantity 25 g
UN Number 3077
Formula Weight 167.21
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Carbazole
EINECSNumber (5)-0150
RTECSNumber FE3150000
TSCA Yes