missing translation for 'onlineSavingsMsg'
Learn More

Carbazole, 96%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 108260010

Catalog No. AC108260010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Carbazole
86-74-8
220°C
95% min. (GC)
C12H9N
1kg
15,1791
Solubility in water: insoluble. Other solubilities: slightly soluble in acetic acid- soluble in h2so4,1g/3mL quinoline- 1g/6mL pyridine- 1g/35mL ether,1g/9mL acetone- 1g/120mL benzene,1g/135mL abs. alcohol- slightly soluble in,petroleum ether and
N1C2=C(C=CC=C2)C2=C1C=CC=C2
167.21
CHEBI:27543
96%
96%
355.0°C
Authentic
Plastic bottle
MFCD00004960
20,433
carbazole, dibenzopyrrole, diphenylenimine, 9-azafluorene, diphenylenimide, diphenyleneimine, dibenzo b,d pyrrole, usaf ek-600, unii-0p2197hhhn
UJOBWOGCFQCDNV-UHFFFAOYSA-N
9H-carbazole
6854
167.21
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Very toxic to aquatic life with long lasting effects.
Causes serious eye irritation.
Suspected of causing cancer.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Avoid breathing dust/fume/

GHS Signal Word: Warning

EINECSNumber : 201-696-0

RTECSNumber : FE3150000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only