missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbazole, 96%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 240.0°C to 246.0°C |
| Color | Beige-Brown or Beige-Yellow |
| Boiling Point | 355.0°C |
| Flash Point | 220°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 95% min. (GC) |
| Packaging | Glass Bottle |
| Quantity | 5 g |
| Beilstein | 20,433 |
| Merck Index | 15,1791 |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement Causes skin irritation. Very toxic to aquatic life with long lasting effects. Causes serious eye irritation. Suspected of causing cancer. May cause respiratory irritation. |
| Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. If skin irritation occurs: Get medical advice/attention. If eye irritation persists: Get medical advice/attention. Avoid breathing dust/fume/ |
| EINECSNumber | 201-696-0 |
| RTECSNumber | FE3150000 |
| TSCA | TSCA |
RUO – Research Use Only