Learn More
Carbazole, 95%
CAS: 86-74-8 | C12H9N | 167.21 g/mol
Supplier: Thermo Scientific Chemicals A1144822
Description
Carbazole and its derivatives are widely used as an intermediate in synthesis of pharmaceuticals, agrochemicals, dyes, pigments and other organic compounds. carbazole is also used in luminescence chemistry as a photosensitizing and additional charge transport material. Carbazole structure is a motif in pharmaceuticals such as carvedilol used to treat high blood pressure and to prevent cardiac arrhythmias and angina.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsCarbazole and its derivatives are widely used as an intermediate in synthesis of pharmaceuticals, agrochemicals, dyes, pigments and other organic compounds. carbazole is also used in luminescence chemistry as a photosensitizing and additional charge transport material. Carbazole structure is a motif in pharmaceuticals such as carvedilol used to treat high blood pressure and to prevent cardiac arrhythmias and angina.
Solubility
It is insoluble in water.
Notes
Stable under ordinary conditions. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
Specifications
| Carbazole | |
| 240°C to 246°C | |
| 354°C to 356°C | |
| Odorless | |
| MFCD00004960 | |
| UN3077 | |
| 14,1790 | |
| It is insoluble in water. | |
| N1C2=C(C=CC=C2)C2=C1C=CC=C2 | |
| 167.21 | |
| CHEBI:27543 | |
| 95% |
| 86-74-8 | |
| 1.15 | |
| 220°C (428°F) | |
| C12H9N | |
| 100 g | |
| 3956 | |
| carbazole, dibenzopyrrole, diphenylenimine, 9-azafluorene, diphenylenimide, diphenyleneimine, dibenzo b,d pyrrole, usaf ek-600, unii-0p2197hhhn | |
| UJOBWOGCFQCDNV-UHFFFAOYSA-N | |
| 9H-carbazole | |
| 6854 | |
| 167.21 |
Chemical Identifiers
| 86-74-8 | |
| 167.21 | |
| UJOBWOGCFQCDNV-UHFFFAOYSA-N | |
| 6854 | |
| 9H-carbazole |
| C12H9N | |
| MFCD00004960 | |
| carbazole, dibenzopyrrole, diphenylenimine, 9-azafluorene, diphenylenimide, diphenyleneimine, dibenzo b,d pyrrole, usaf ek-600, unii-0p2197hhhn | |
| CHEBI:27543 | |
| N1C2=C(C=CC=C2)C2=C1C=CC=C2 |
Safety and Handling
GHS H Statement
H351
Suspected of causing cancer.
P201-P202-P261-P264b-P271-P280i-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P332+P313-P362-P501c
H315-H319-H335-H351
DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber : 201-696-0
RTECSNumber : FE3150000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only