missing translation for 'onlineSavingsMsg'
Learn More

Carbazole, 95%

Catalog No. AAA1144818
Change view
Click to view available options
Quantity:
50 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1144818 50 g
AAA1144822 100 g
AAA1144836 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1144818 Supplier Thermo Scientific Chemicals Supplier No. A1144818
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 86-74-8 | C12H9N | 167.21 g/mol

Carbazole and its derivatives are widely used as an intermediate in synthesis of pharmaceuticals, agrochemicals, dyes, pigments and other organic compounds. carbazole is also used in luminescence chemistry as a photosensitizing and additional charge transport material. Carbazole structure is a motif in pharmaceuticals such as carvedilol used to treat high blood pressure and to prevent cardiac arrhythmias and angina.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Carbazole and its derivatives are widely used as an intermediate in synthesis of pharmaceuticals, agrochemicals, dyes, pigments and other organic compounds. carbazole is also used in luminescence chemistry as a photosensitizing and additional charge transport material. Carbazole structure is a motif in pharmaceuticals such as carvedilol used to treat high blood pressure and to prevent cardiac arrhythmias and angina.

Solubility
It is insoluble in water.

Notes
Stable under ordinary conditions. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.

Chemical Identifiers

CAS 86-74-8
Molecular Formula C12H9N
Molecular Weight (g/mol) 167.21
MDL Number MFCD00004960
InChI Key UJOBWOGCFQCDNV-UHFFFAOYSA-N
Synonym carbazole, dibenzopyrrole, diphenylenimine, 9-azafluorene, diphenylenimide, diphenyleneimine, dibenzo b,d pyrrole, usaf ek-600, unii-0p2197hhhn
PubChem CID 6854
ChEBI CHEBI:27543
IUPAC Name 9H-carbazole
SMILES N1C2=C(C=CC=C2)C2=C1C=CC=C2

Specifications

Melting Point 240°C to 246°C
Density 1.15
Boiling Point 354°C to 356°C
Flash Point 220°C (428°F)
Odor Odorless
Quantity 50 g
UN Number UN3077
Beilstein 3956
Merck Index 14,1790
Solubility Information It is insoluble in water.
Formula Weight 167.21
Percent Purity 95%
Chemical Name or Material Carbazole
Show More Show Less
Hazard Category H315-H319-H335-H351
Hazard Statement GHS H Statement
H351
Suspected of causing cancer.
Precautionary Statement P201-P202-P261-P264b-P271-P280i-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 201-696-0
RTECSNumber FE3150000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only