missing translation for 'onlineSavingsMsg'
Learn More

Carbazole-2-boronic acid pinacol ester, tech. 90%, Thermo Scientific™

$1046.82 - $1046.82

Chemical Identifiers

CAS 871125-67-6
Molecular Formula C18H20BNO2
Molecular Weight (g/mol) 293.173
MDL Number MFCD07784368
InChI Key RLSJGSFDSSYNPL-UHFFFAOYSA-N
Synonym 9h-carbazole-2-boronic acid pinacol ester, 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-9h-carbazole, 9h-carbazole,4a,9a-dihydro-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, carbazole-2-boronic acid pinacol ester, 9h-carbazol-2-ylboronic acid pinacol ester, 9h-carbazol-2-yl boronic acid pinacol ester, 2-tetramethyl-1,3,2-dioxaborolan-2-yl-9h-carbazole, 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl-9h-carbazole, 2-9h-carbazol-2-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
PubChem CID 46738013
IUPAC Name 2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-carbazole
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC3=C(C=C2)C4=CC=CC=C4N3
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5548403
View Documents
Thermo Scientific Chemicals
H5548403
1 g
Each for $1,046.82
Only null left
 
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

871125-67-6
293.173
RLSJGSFDSSYNPL-UHFFFAOYSA-N
46738013
B1(OC(C(O1)(C)C)(C)C)C2=CC3=C(C=C2)C4=CC=CC=C4N3
C18H20BNO2
MFCD07784368
9h-carbazole-2-boronic acid pinacol ester, 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-9h-carbazole, 9h-carbazole,4a,9a-dihydro-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, carbazole-2-boronic acid pinacol ester, 9h-carbazol-2-ylboronic acid pinacol ester, 9h-carbazol-2-yl boronic acid pinacol ester, 2-tetramethyl-1,3,2-dioxaborolan-2-yl-9h-carbazole, 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl-9h-carbazole, 2-9h-carbazol-2-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-carbazole

Specifications

871125-67-6
MFCD07784368
9h-carbazole-2-boronic acid pinacol ester, 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-9h-carbazole, 9h-carbazole,4a,9a-dihydro-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, carbazole-2-boronic acid pinacol ester, 9h-carbazol-2-ylboronic acid pinacol ester, 9h-carbazol-2-yl boronic acid pinacol ester, 2-tetramethyl-1,3,2-dioxaborolan-2-yl-9h-carbazole, 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl-9h-carbazole, 2-9h-carbazol-2-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
B1(OC(C(O1)(C)C)(C)C)C2=CC3=C(C=C2)C4=CC=CC=C4N3
293.173
293.17
Technical
Carbazole-2-boronic acid pinacol ester
C18H20BNO2
1 g
RLSJGSFDSSYNPL-UHFFFAOYSA-N
2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-carbazole
46738013
90%
Solid
Videos
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'