missing translation for 'onlineSavingsMsg'
Learn More

Carbamylcholine Chloride 99.0+%, TCI America™

Catalog No. C059610G
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C059610G 10 g
C059625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C059610G Supplier TCI America Supplier No. C059610G

Chemical Identifiers

CAS 51-83-2
Molecular Formula C6H15ClN2O2
Molecular Weight (g/mol) 182.65
MDL Number MFCD00012011
InChI Key AIXAANGOTKPUOY-UHFFFAOYSA-N
Synonym carbachol, carbamoylcholine chloride, carbachol chloride, carbacholine, carbocholine, jestryl, doryl, carbamylcholine chloride, miostat, isopto carbachol
PubChem CID 5831
ChEBI CHEBI:3385
IUPAC Name 2-(trimethylazaniumyl)ethyl carbamate chloride
SMILES [Cl-].C[N+](C)(C)CCOC(N)=O

Specifications

Melting Point 189°C
Color White
Quantity 10 g
UN Number 2811
Formula Weight 182.65
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Carbamylcholine Chloride
RTECSNumber GA0875000
TSCA Yes