missing translation for 'onlineSavingsMsg'
Learn More

Carbamylcholine chloride, 98+%

Catalog No. p-4228989
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC108240050 5 g Glass bottle
AC108240250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC108240050 Supplier Thermo Scientific Chemicals Supplier No. 108240050
Only null left

CAS: 51-83-2 | C6H15ClN2O2 | 182.65 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 51-83-2
Molecular Formula C6H15ClN2O2
Molecular Weight (g/mol) 182.65
MDL Number MFCD00012011
InChI Key AIXAANGOTKPUOY-UHFFFAOYSA-N
Synonym carbachol, carbamoylcholine chloride, carbachol chloride, carbacholine, carbocholine, jestryl, doryl, carbamylcholine chloride, miostat, isopto carbachol
PubChem CID 5831
ChEBI CHEBI:3385
IUPAC Name 2-carbamoyloxyethyl(trimethyl)azanium;chloride
SMILES [Cl-].C[N+](C)(C)CCOC(N)=O

Specifications

Melting Point 200°C to 204°C
Color White
Physical Form Crystalline Powder
Quantity 5 g
Assay Percent Range 99%
Linear Formula H2NCO2CH2CH2N(CH3)3Cl
Merck Index 15,1781
Solubility Information Solubility in water: 1g/mL. Other solubilities: 1g/50 mL alcohol,1g/10 mL methanol,practically insoluble in chloroform and ether
Formula Weight 182.65
Percent Purity 99%
Infrared Spectrum Authentic
Loss on Drying 2.0% max. (105°C, 2 hrs)
Packaging Glass bottle
Chemical Name or Material Carbamylcholine chloride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Fatal if swallowed.
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wash face,hands and any exposed skin thoroughly after handling.
EINECSNumber 200-127-3

RUO – Research Use Only