missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbamylcholine chloride, 98+%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 200°C to 204°C |
| Color | White |
| Physical Form | Crystalline Powder |
| Quantity | 25 g |
| Infrared Spectrum | Authentic |
| Loss on Drying | 2.0% max. (105°C, 2 hrs) |
| Packaging | Glass bottle |
| Assay Percent Range | 99% |
| Linear Formula | H2NCO2CH2CH2N(CH3)3Cl |
| Merck Index | 15,1781 |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Fatal if swallowed. |
| Precautionary Statement | GHS P Statement IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. Wash face,hands and any exposed skin thoroughly after handling. |
| EINECSNumber | 200-127-3 |
RUO – Research Use Only