missing translation for 'onlineSavingsMsg'
Learn More

Carbamoyl-2,2,5,5-tetramethyl-3-pyrrolin-1-yloxy, free radical, 99%

Catalog No. AAA1738106
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1738106 5 g
AAA1738103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1738106 Supplier Thermo Scientific Chemicals Supplier No. A1738106
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3229-73-0 | C9H16N2O2 | 184.24 g/mol

It is employed as a highly reactive spin-label.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a highly reactive spin-label.

Solubility
Soluble in acetone, chloroform, dichloromethane, ethyl acetate and methanol.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 3229-73-0
Molecular Formula C9H16N2O2
Molecular Weight (g/mol) 184.24
MDL Number MFCD00005341
InChI Key GMXOTWPDUBXCMS-UHFFFAOYSA-N
Synonym 2,2,5,5-Tetramethyl-3-pyrrolin-1-oxyl-3-carboxamide
IUPAC Name 1-hydroxy-2,2,5,5-tetramethyl-2,5-dihydro-1H-pyrrole-3-carboxamide
SMILES CC1(C)C=C(C(N)=O)C(C)(C)N1O

Specifications

Melting Point 198°C to 200°C
Quantity 5 g
Beilstein 4138020
Solubility Information Soluble in acetone,chloroform,dichloromethane,ethyl acetate and methanol.
Formula Weight 183.23
Percent Purity 99%
Chemical Name or Material Carbamoyl-2,2,5,5-tetramethyl-3-pyrrolin-1-yloxy, free radical
EINECSNumber 221-765-9
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only