missing translation for 'onlineSavingsMsg'
Learn More

Carbachol, 98.3%

Catalog No. AAL0667414
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0667414 25 g
AAL0667403 1 g
AAL0667406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0667414 Supplier Thermo Scientific Chemicals Supplier No. L0667414

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Carbachol, CAS # 51-83-2, is a parasympathomimetic that acts on acetylcholine receptors. | CAS: 51-83-2 | C6H15ClN2O2 | 182.65 g/mol

Carbachol is a cholinergic agonist and is used for the treatment of glaucoma. It is also used as ophthalmic solution (eye drops) during ophthalmic surgery.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Carbachol is an ammonium salt and a choline ester that acts on both muscarinic and nicotinic receptors
  • Carbachol activates the sympathetic nervous system; it leads to intracellular increases in Ca2+ and activates associated signaling pathways
  • Carbachol is not degraded by the cholinesterases

Application

  • Carbachol can inhibit apoptotic cell death, making it useful for the study of cell survival and growth in laboratory settings
  • Carbachol can be used for laboratory research of any biological pathway that depends on acetylcholine, including sleep cycles and motor control

Chemical Identifiers

CAS 51-83-2
Molecular Formula C6H15ClN2O2
Molecular Weight (g/mol) 182.65
MDL Number MFCD00012011
InChI Key AIXAANGOTKPUOY-UHFFFAOYSA-N
Synonym carbachol, carbamoylcholine chloride, carbachol chloride, carbacholine, carbocholine, jestryl, doryl, carbamylcholine chloride, miostat, isopto carbachol
PubChem CID 5831
ChEBI CHEBI:3385
IUPAC Name 2-(trimethylazaniumyl)ethyl carbamate chloride
SMILES [Cl-].C[N+](C)(C)CCOC(N)=O

Specifications

Melting Point ∽210°C (decomposition)
Quantity 25 g
UN Number UN2811
Beilstein 3917459
Sensitivity Hygroscopic
Merck Index 14,1779
Solubility Information Soluble in water and dimethyl sulfoxide.
Formula Weight 182.65
Percent Purity 98.3%
Chemical Name or Material Carbachol
Hazard Category H300-H500
Hazard Statement GHS H Statement
H300
Fatal if swallowed.
Precautionary Statement P264b-P270-P301+P310-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 200-127-3
RTECSNumber GA0875000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only