missing translation for 'onlineSavingsMsg'
Learn More

Carazolol 98.0+%, TCI America™

Catalog No. p-7125538
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C2578200MG 200 mg
C25781G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C2578200MG Supplier TCI America Supplier No. C2578200MG
Only null left

Chemical Identifiers

CAS 57775-29-8
Molecular Formula C18H22N2O2
Molecular Weight (g/mol) 298.386
MDL Number MFCD00864588
InChI Key BQXQGZPYHWWCEB-UHFFFAOYSA-N
Synonym 1-(9H-Carbazol-4-yloxy)-3-(isopropylamino)-2-propanol
PubChem CID 71739
IUPAC Name 1-(9H-carbazol-4-yloxy)-3-(propan-2-ylamino)propan-2-ol
SMILES CC(C)NCC(COC1=CC=CC2=C1C3=CC=CC=C3N2)O

Specifications

Melting Point 136°C
Color White-Yellow
Quantity 200 mg
UN Number 2811
Formula Weight 298.39
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Carazolol
RTECSNumber UA8685000
TSCA No