missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Caramel Color, Food Grade, Spectrum™ Chemical
Description
Spectrum™ Chemical Caramel Color, Food Grade
Specifications
Specifications
| CAS Max % | 100% |
| Packaging | Amber Glass Bottle |
| Quantity | 500 mL |
| Specific Gravity | 1.315-1.325 |
| Grade | Food Grade |