missing translation for 'onlineSavingsMsg'
Learn More

CleanSpace™ PAF-1024 Car Charger

Catalog No. 17800627
Change view
Click to view available options
For Use With (Equipment):
CleanSpace2™
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
17800627 CleanSpace2™
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17800627 Supplier CleanSpace™ Supplier No. PAF1024

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Car Charger designed for use with all CleanSpace2™ PAPR models.

  • Black
  • For use with all CleanSpace2™ Respirator

Specifications

Color Black
Product Type Car Charger
For Use With (Equipment) CleanSpace2™
Model PAF-1024