missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ CaptureSelect™ LambdaXP RoboColumn™ Pre-packed Columns

Catalog No. 5943752200
Change view
Click to view available options
Bed Volume:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Bed Volume
5943752200 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 5943752200 Supplier Thermo Scientific™ Supplier No. 5943752200

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CaptureSelect RoboColumns are small chromatography columns used for fully automated and parallel chromatographic separations on robotic liquid handling workstations.

CaptureSelect RoboColumn™ are small chromatography columns used for fully automated and parallel chromatographic separations on robotic liquid handling workstations.

The CaptureSelect LambdaXP affinity matrix in this RoboColumn recognizes the constant domain of the human lambda light chain enabling purification of all IgG isotypes, including IgG, IgA, IgM, IgD, and IgE, directly from complex source materials in a robust and efficient purification process with excellent purity obtained in a single step.

Features of the LambdaXP affinity matrix include:

  • Efficient elution of Fab fragments and immunoglobulins containing a lambda light chain; no cross binding to the Fc-region or kappa light chains
  • One-step purification from crude material with high purity and yield
  • Human specific
  • Non-animal-derived

Free of animal components

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

Characteristics

  • Column size: 200 μL, 1 strip, 8 columns (8-row)
  • Column dimension: 5-mm diameter, 10-mm bed height
  • Matrix: agarose-based, epoxide-activated
  • Average particle size: 65 μm
  • Ligand:CaptureSelect LambdaXP affinity ligand
  • Ligand coupling: epoxide coupling
  • Binding capacity: >30 g IgG/L resin depending on flow rate, column height, and contact time
  • Elution conditions: 25 mM sodium acetate, pH 3.5
  • Formulation buffer: 20% (v/v) ethanol
  • Operating pressure: 2 bar (maximum)
  • Flow rate: 16–150 cm/hr

RoboColumn is a trademark of Repligen.

Specifications

Max. Pressure 2 bar
Pore Size Not Applicable
Column Capacity 200 μL
Length (Metric) 1 cm
Quantity 1 Strip, 8 Columns
Particle Size 65 μm
For Use With (Equipment) Robotic Fluid Handlers
Type Affinity Column
Diameter (Metric) Inner 0.5 cm
Content And Storage Store at 2°C to 8°C. Protect from light. Do not freeze.
Product Line CaptureSelect
Ligand Type CaptureSelect LambdaXP Affinity Ligand
Matrix Agarose
Shipping Condition Wet Ice
Column Format Pre-packed RoboColumn
Bed Volume 200 μL
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.